4-Difluoromethoxy-3-hydroxybenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Difluoromethoxy)-3-hydroxybenzaldehyde

074041-5G
102534-322EA 300 USD
102534-322
4-Difluoromethoxy-3-hydroxybenzaldehyde ≥95%
4-Difluoromethoxy-3-hydroxybenzaldehyde

Hazard: Irritant

Formula: C₈H₆F₂O₃
MW: 188.1283 g/mol
MDL Number: MFCD04406687
CAS Number: 151103-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR