4-Bromo-5-chloroanthranilic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-bromo-5-chlorobenzoic acid

074040-5G
102534-316EA 307.69 USD
102534-316
4-Bromo-5-chloroanthranilic acid ≥95%
4-Bromo-5-chloroanthranilic acid

Hazard: Irritant

Formula: C₇H₅BrClNO₂
MW: 250.4800 g/mol
MDL Number: MFCD10000554
CAS Number: 150812-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR