1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥95%

Supplier: Matrix Scientific

Synonyms: Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)

074028-5G
102534-252EA 96.92 USD
102534-252
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥95%
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride

Hazard: Irritant

Formula: C₂₆H₂₄Cl₂NiP₂
MDL Number: MFCD00013313
CAS Number: 14647-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR