3-Fluoro-5-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

074022-1G 074022-5G
102534-224EA 38.46 USD
102534-224 102534-226
3-Fluoro-5-nitrobenzoic acid ≥95%
3-Fluoro-5-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
MDL Number: MFCD05864360
CAS Number: 14027-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR