3-Bromo-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

074014-5G
102534-186EA 184.62 USD
102534-186
3-Bromo-2-methoxypyridine ≥95%
3-Bromo-2-methoxypyridine

Hazard: Irritant/Light Sensitive/Moisture Sensitive/Stored under Argon

Formula: C₆H₆BrNO
Boiling Pt: 70…73 °C (3 mmHg)
Density: 1.58
Flash Pt: 93.3 °C
MDL Number: MFCD03095349
CAS Number: 13472-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR