2-Bromo-5-phenyloxazole ≥95%
Supplier: Matrix Scientific
074008-1G
074008-5G
074008-250MG
102534-150EA
456.92
USD
102534-150
102534-152
102534-154
2-Bromo-5-phenyloxazole ≥95%
2-Bromo-5-phenyloxazole
Hazard: Irritant
Formula:
C₉H₆BrNO MW: 224.0540 g/mol |
MDL Number:
MFCD12828094 CAS Number: 129053-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...