5-Bromoisoindoline hydrochloride ≥95%

Supplier: Matrix Scientific

074005-250MG 074005-1G
102534-134EA 315.38 USD
102534-134 102534-136
5-Bromoisoindoline hydrochloride ≥95%
5-Bromoisoindoline hydrochloride

Hazard: Irritant

Formula: C₈H₉BrClN
MW: 234.5200 g/mol
MDL Number: MFCD10700135
CAS Number: 919346-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR