5-Bromo-m-toluonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-5-methylbenzonitrile
074002-1G
074002-25G
074002-5G
102534-120EA
221.54
USD
102534-120
102663-968
102534-118
5-Bromo-m-toluonitrile ≥95%
5-Bromo-m-toluonitrile
Hazard: Irritant
Formula:
C₈H₆BrN MW: 196.0400 g/mol |
MDL Number:
MFCD08061970 CAS Number: 124289-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...