2,5-Dimethylpyrazine ≥95%
Supplier: Matrix Scientific
074000-100G
074000-10G
102534-110EA
492.31
USD
102534-110
102534-108
2,5-Dimethylpyrazine ≥95%
2,5-Dimethylpyrazine
Hazard: Irritant
Formula:
C₆H₈N₂ MW: 108.14 g/mol |
MDL Number:
MFCD00006147 CAS Number: 123-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...