1-(3-Chloropropyl)-4-methylpiperazine ≥95%

Supplier: Matrix Scientific

073968-1G
102533-952EA 516.92 USD
102533-952
1-(3-Chloropropyl)-4-methylpiperazine ≥95%
1-(3-Chloropropyl)-4-methylpiperazine

Hazard: Irritant

Formula: C₈H₁₇ClN₂
Boiling Pt: 243 °C
Density: 1.015
Flash Pt: 101 °C
CAS Number: 104-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR