3-Amino-6-bromo-4-methylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Amino-2-bromo-4-methylpyridine
073951-5G
102533-866EA
255.38
USD
102533-866
3-Amino-6-bromo-4-methylpyridine ≥95%
3-Amino-6-bromo-4-methylpyridine
Hazard: Irritant
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol |
MDL Number:
MFCD08277280 CAS Number: 156118-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...