2-Chloroquinolin-4-ol ≥95%
Supplier: Matrix Scientific
073950-250MG
073950-1G
102533-860EA
275.38
USD
102533-860
102533-862
2-Chloroquinolin-4-ol ≥95%
2-Chloroquinolin-4-ol
Hazard: Irritant
Formula:
C₉H₆ClNO MW: 179.6030 g/mol |
MDL Number:
MFCD08705676 CAS Number: 771555-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...