4-Chlorofuro[3,2-c]pyridine ≥95%

Supplier: Matrix Scientific

073947-5G 073947-1G
102533-846EA 276.92 USD
102533-846 102533-844
4-Chlorofuro[3,2-c]pyridine ≥95%
4-Chlorofuro[3,2-c]pyridine

Hazard: Irritant

Formula: C₇H₄ClNO
MW: 153.5658 g/mol
MDL Number: MFCD02179768
CAS Number: 31270-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR