tert-Butyl-4-hydroxy-2-oxopiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-hydroxy-2-oxopiperidine-1-carboxylate

073917-5G 073917-1G 073917-250MG
102533-690EA 640 USD
102533-690 102533-694 102533-692
tert-Butyl-4-hydroxy-2-oxopiperidine-1-carboxylate ≥95%
tert-Butyl-4-hydroxy-2-oxopiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MW: 215.2500 g/mol
MDL Number: MFCD16659601
CAS Number: 1245646-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR