2-Chloro-5-pyridinesulfonyl chloride ≥95%

Supplier: Matrix Scientific

073903-5G 073903-1G 073903-25G
102533-618EA 129.23 USD
102533-618 102533-616 102833-658
2-Chloro-5-pyridinesulfonyl chloride ≥95%
2-Chloro-5-pyridinesulfonyl chloride

Hazard: Irritant

Formula: C₅H₃Cl₂NO₂S
MW: 212.0538 g/mol
MDL Number: MFCD03541049
CAS Number: 6684-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR