2,6-Dichloro-3-fluoropyridine ≥95%
Supplier: Matrix Scientific
073892-5G
073892-1G
102533-556EA
44.62
USD
102533-556
102533-558
2,6-Dichloro-3-fluoropyridine ≥95%
2,6-Dichloro-3-fluoropyridine
Hazard: Irritant
Formula:
C₅H₂Cl₂FN MW: 165.9805 g/mol |
MDL Number:
MFCD09907815 CAS Number: 52208-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...