tert-Butyl pyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: N-BOC-Pyrrolidinetert-butyl pyrrolidine-1-carboxylatepyrrolidine-1-carboxylic acid tert-butyl ester

073887-5G
102533-532EA 32.31 USD
102533-532
tert-Butyl pyrrolidine-1-carboxylate ≥95%
tert-Butyl pyrrolidine-1-carboxylate

Hazard: Irritant/Store in Freezer

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Boiling Pt: 130 °C (20 mmHg)
Density: 0.977
MDL Number: MFCD00216581
CAS Number: 86953-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR