1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene ≥95%

Supplier: Matrix Scientific

073885-25G
102663-958EA 126.15 USD
102663-958
1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene ≥95%
1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene

Hazard: Irritant

Formula: C₇H₃BrF₄O
MW: 259.00 g/mol
MDL Number: MFCD06657548
CAS Number: 168971-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR