4-Methoxy-2-nitrobenzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methoxy-2-nitrobenzene-1-sulphonyl chloride4-Methoxy-2-nitrobenzene-1-sulfonyl chloride

073857-5G 073857-10G 073857-1G
102533-372EA 220 USD
102533-372 102533-370 102533-368
4-Methoxy-2-nitrobenzenesulfonyl chloride ≥95%
4-Methoxy-2-nitrobenzenesulphonyl chloride

Hazard: Irritant

Formula: C₇H₆ClNO₅S
MW: 251.65 g/mol
MDL Number: MFCD00143333
CAS Number: 18092-54-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR