2-Amino-6-chloro-5-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-5-methylpyridin-2-amine

073781-250MG 073781-1G 073781-5G
102531-970EA 3709.23 USD
102531-970 102531-968 102531-966
2-Amino-6-chloro-5-methylpyridine ≥95%
2-Amino-6-chloro-5-methylpyridine

Hazard: Irritant

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
MDL Number: MFCD13193513
CAS Number: 442129-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR