2-Amino-3-chlorobenzylalkohol ≥95%
Supplier: Matrix Scientific
Synonyms:
(2-Amino-3-chlorophenyl)methanol
073755-1G
073755-5G
102531-832EA
1473.85
USD
102531-832
102531-830
2-Amino-3-chlorobenzylalkohol ≥95%
2-Amino-3-chlorobenzylalkohol
Hazard: Irritant
Formula:
C₇H₈ClNO MW: 157.6000 g/mol |
MDL Number:
MFCD13193432 CAS Number: 61487-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...