4,4'-Dimethoxytrityl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4,4'-Dimethoxytriphenylmethyl chlorideDMT-Cl

073653-100G
102531-302EA 130.77 USD
102531-302
4,4'-Dimethoxytrityl chloride ≥95%
4,4'-Dimethoxytrityl chloride

Hazard: Irritant/Moisture Sensitive

Formula: C₂₁H₁₉ClO₂
MW: 338.8 g/mol
Melting Pt: 121…127 °C
MDL Number: MFCD00008409
CAS Number: 40615-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR