5-Bromoindol-3-ylacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromoindole-3-acetic acid2-(5-Bromo-1H-indol-3-yl)acetic acid

073652-1G 073652-5G
102531-298EA 860 USD
102531-298 102531-296
5-Bromoindol-3-ylacetic acid ≥95%
5-Bromoindol-3-ylacetic acid

Hazard: Irritant

Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
MDL Number: MFCD00005637
CAS Number: 40432-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR