Ethyl-3-hydroxy-8,9-dihydro-5H-pyridazino[3,4-d]azepine-7(6H)-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-hydroxy-8,9-dihydro-5H-pyridazino[3,4-d]azepine-7(6H)-carboxylate

074522-1G
102536-862EA 2610.77 USD
102536-862
Ethyl-3-hydroxy-8,9-dihydro-5H-pyridazino[3,4-d]azepine-7(6H)-carboxylate ≥95%
Ethyl-3-hydroxy-8,9-dihydro-5H-pyridazino[3,4-d]azepine-7(6H)-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₃
MW: 237.2600 g/mol
MDL Number: MFCD12756173
CAS Number: 1190897-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR