4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid ethyl ester2-[(4-Ethoxycarbonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane4-(Ethoxycarbonylmethyl)phenylboronic acid Pinacol esterEthyl 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetateEthyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate

074434-1G
102536-412EA 67.85 USD
102536-412
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester ≥95%
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₆H₂₃BO₄
MW: 290.1624 g/mol
MDL Number: MFCD08704684
CAS Number: 859169-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR