6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Isoxepac2-(11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic acid6H-11-Oxodibenzo[b,e]oxepin-2-acetic Acid

074300-5G
102535-704EA 463.08 USD
102535-704
6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic acid ≥95%
6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic acid

Hazard: Irritant

Formula: C₁₆H₁₂O₄
MW: 268.27 g/mol
MDL Number: MFCD00242952
CAS Number: 55453-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR