5-(Bromomethyl)-1-methyl-1H-benzo[d][1,2,3]triazole ≥95%

Supplier: Matrix Scientific

074263-1G
102535-510EA 1383.08 USD
102535-510
5-(Bromomethyl)-1-methyl-1H-benzo[d][1,2,3]triazole ≥95%
5-(Bromomethyl)-1-methyl-1H-benzo[d][1,2,3]triazole

Hazard: Irritant

Formula: C₈H₈BrN₃
CAS Number: 499770-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR