tert-Butyl-2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate
074112-1G
074112-250MG
102534-706EA
710.77
USD
102534-706
102534-708
tert-Butyl-2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate ≥95%
tert-Butyl-2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₀N₂O₃ MW: 288.3400 g/mol |
MDL Number:
MFCD08458288 CAS Number: 205383-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...