7-Nitro-2,3-dihydro-1,4-benzodioxine-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
7-Nitro-1,4-benzodioxane-6-carboxylic Acid
073941-1G
073941-250MG
102533-816EA
400
USD
102533-816
102533-818
7-Nitro-2,3-dihydro-1,4-benzodioxine-6-carboxylic acid ≥95%
7-Nitro-2,3-dihydro-1,4-benzodioxine-6-carboxylic acid
Hazard: Irritant
Formula:
C₉H₇NO₆ MW: 225.1550 g/mol |
MDL Number:
MFCD01116988 CAS Number: 57672-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...