2-Chloro-3-fluorophenol ≥95%

Supplier: Matrix Scientific

073613-5G
102531-094EA 192.31 USD
102531-094
2-Chloro-3-fluorophenol ≥95%
2-Chloro-3-fluorophenol

Hazard: Irritant

Formula: C₆H₄ClFO
MW: 146.5500 g/mol
MDL Number: MFCD08166413
CAS Number: 863870-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR