6-Nitroquinoline ≥95%

Supplier: Matrix Scientific

073600-100G
102531-026EA 239.2 USD
102531-026
6-Nitroquinoline ≥95%
6-Nitroquinoline

Hazard: Irritant

Formula: C₉H₆N₂O₂
MDL Number: MFCD00006799
CAS Number: 613-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR