4-Bromotetrahydropyran ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromotetrahydro-2H-pyran4-Bromooxane

073578-5G
102530-916EA 123.08 USD
102530-916
4-Bromotetrahydropyran ≥95%
4-Bromotetrahydropyran

Hazard: Irritant

Formula: C₅H₉BrO
MW: 165.0300 g/mol
MDL Number: MFCD07371501
CAS Number: 25637-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR