(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
H-D-HoPro-OMe.HCl
, H-D-Homopro-OMe·HCl
073544-5G
073544-1G
102530-728EA
152.31
USD
102530-728
102530-730
(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride ≥95%
(R)-Piperidine-2-carboxylic acid methyl ester hydrochloride
Hazard: Irritant
Formula:
C₇H₁₄ClNO₂ MW: 179.644 g/mol |
MDL Number:
MFCD00237412 CAS Number: 18650-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...