5-Amino-2,3-dichloropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-5,6-dichloropyridine2,3-Dichloropyridin-5-amine2,3-Dichloro-5-pyridylamine

073537-5G
102530-694EA 236.92 USD
102530-694
5-Amino-2,3-dichloropyridine ≥95%
5-Amino-2,3-dichloropyridine

Hazard: Irritant

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
MDL Number: MFCD03840434
CAS Number: 98121-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR