2-Amino-4-methoxy-6-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073523-100G
102530-620EA 484.62 USD
102530-620
2-Amino-4-methoxy-6-methylpyrimidine ≥95%
2-Amino-4-methoxy-6-methylpyrimidine

Hazard: Irritant

Formula: C₆H₉N₃O
MDL Number: MFCD00059768
CAS Number: 7749-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR