(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride

073520-10G 073520-100G
102530-604EA 178.46 USD
102530-604 102530-606
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥95%
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₂
MW: 213.6600 g/mol
MDL Number: MFCD00800357
CAS Number: 77497-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR