Histamine dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride4-(2-Aminoethyl)-1H-imidazole dihydrochloride

073512-100G
102530-562EA 141.54 USD
102530-562
Histamine dihydrochloride ≥95%
Histamine dihydrochloride

Hazard: Irritant

Formula: C₅H₁₁Cl₂N₃
MW: 184.07 g/mol
Melting Pt: 245…249 °C
MDL Number: MFCD00012703
CAS Number: 56-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR