4-Chloro-2-nitrobenzaldehyde ≥95%
Supplier: Matrix Scientific
073507-1G
073507-5G
073507-10G
102530-536EA
55.38
USD
102530-536
102530-538
102530-540
4-Chloro-2-nitrobenzaldehyde ≥95%
4-Chloro-2-nitrobenzaldehyde
Hazard: Irritant
Formula:
C₇H₄ClNO₃ MW: 185.57 g/mol Melting Pt: 67…68 °C |
MDL Number:
MFCD00056104 CAS Number: 5551-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...