5-Methoxy-1-indanone ≥95%

Supplier: Matrix Scientific

073492-5G 073492-25G
102530-452EA 94.3 USD
102530-452 102530-454
5-Methoxy-1-indanone ≥95%
5-Methoxy-1-indanone

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00003789
CAS Number: 5111-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR