Homopiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: Perhydro-1,4-diazepine1,4-Diazepane1,4-Diazacycloheptane

073491-10G
102530-448EA 15.38 USD
102530-448
Homopiperazine ≥95%
Homopiperazine

Hazard: Corrosive/Air Sensitive/Hygroscopic

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 168…170 °C
Melting Pt: 38…44 °C
Density: 0.95
MDL Number: MFCD00006933
CAS Number: 505-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR