1,4'-Bipiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Piperidinopiperidine

073487-5G
102530-430EA 29.23 USD
102530-430
1,4'-Bipiperidine ≥95%
1,4'-Bipiperidine

Hazard: Irritant

Formula: C₁₀H₂₀N₂
MW: 168.28 g/mol
MDL Number: MFCD00006475
CAS Number: 4897-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR