4-Bromoquinazoline ≥95%
Supplier: Matrix Scientific
073469-250MG
073469-1G
102530-336EA
1473.85
USD
102530-336
102530-334
4-Bromoquinazoline ≥95%
4-Bromoquinazoline
Hazard: Irritant
Formula:
C₈H₅BrN₂ MW: 209.04 g/mol |
MDL Number:
MFCD10697879 CAS Number: 354574-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...