(1R,2S)-(-)-2-Amino-1,2-diphenylethanol ≥95%

Supplier: Matrix Scientific

073441-5G
102530-186EA 92.31 USD
102530-186
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol ≥95%
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol

Hazard: Irritant

Formula: C₁₄H₁₅NO
MDL Number: MFCD00074960
CAS Number: 23190-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR