2-Amino-3-nitro-5-fluoropyridine ≥95%
Supplier: Matrix Scientific
073426-5G
073426-1G
102530-098EA
533.85
USD
102530-098
102530-100
2-Amino-3-nitro-5-fluoropyridine ≥95%
2-Amino-3-nitro-5-fluoropyridine
Hazard: Irritant
Formula:
C₅H₄FN₃O₂ MW: 157.1 g/mol |
MDL Number:
MFCD04972413 CAS Number: 212268-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...