5-Nitro-2-picoline ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-5-nitropyridine

073425-5G 073425-10G 073425-1G
102530-092EA 161.54 USD
102530-092 102530-094 102530-096
5-Nitro-2-picoline ≥95%
5-Nitro-2-picoline

Hazard: Irritant

Formula: C₆H₆N₂O₂
MW: 138.12 g/mol
MDL Number: MFCD04114179
CAS Number: 21203-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR