3-Methoxypyridazine ≥95%

Supplier: Matrix Scientific

073423-1G 073423-5G
102530-082EA 533.85 USD
102530-082 102530-084
3-Methoxypyridazine ≥95%
3-Methoxypyridazine

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.1139 g/mol
MDL Number: MFCD00234098
CAS Number: 19064-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR