3-Amino-2-methoxy-6-picoline ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-2-methoxy-6-methylpyridine
073421-10G
073421-1G
073421-5G
102530-070EA
427.69
USD
102530-070
102530-072
102530-074
3-Amino-2-methoxy-6-picoline ≥95%
3-Amino-2-methoxy-6-picoline
Hazard: Irritant
Formula:
C₇H₁₀N₂O |
MDL Number:
MFCD03095079 CAS Number: 186413-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...