4-Chloroisoquinoline ≥95%

Supplier: Matrix Scientific

073410-250MG 073410-5G 073410-1G
102530-018EA 250.11 USD
102530-018 102530-020 102530-022
4-Chloroisoquinoline ≥95%
4-Chloroisoquinoline

Hazard: Irritant

Formula: C₉H₆ClN
MW: 163.6036 g/mol
MDL Number: MFCD00234490
CAS Number: 1532-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR