Quinoline-2-ethanol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(2-Hydroxyethyl)quinoline
073309-250MG
073309-1G
102529-488EA
684.62
USD
102529-488
102529-490
Quinoline-2-ethanol ≥95%
Quinoline-2-ethanol
Hazard: Irritant
Formula:
C₁₁H₁₁NO MW: 173.21 g/mol |
MDL Number:
MFCD01685503 CAS Number: 1011-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...