Methyl-4-bromo-2-methylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-bromo-2-methylbenzoate

073278-5G
102529-328EA 58.46 USD
102529-328
Methyl-4-bromo-2-methylbenzoate ≥95%
Methyl 4-bromo-2-methylbenzoate

Hazard: Irritant

Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
MDL Number: MFCD09954957
CAS Number: 99548-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR